Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranin O, 1% AQ, Vion Biosciences
SCI_ED

Catalog No. S02487 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02487 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02487 Supplier Aldon Corporation Supplier No. VNSS0030500ML
Add to Cart
Add to Cart

Histology grade 1% aqueous safranin O is an effective stain for visualizing cell nuclei and as a counterstain in Gram staining.

  • Histology grade 1% aqueous safranin O is an effective stain for visualizing cell nuclei and as a counterstain in Gram staining.

Specifications

Boiling Point ~100°C (212°F in water)
CAS 7732-18-5
Color Dark Red-Brown
Concentration 1%
Description Dark red-brown liquid with no odor.
Detection Method Safranin O staining of cartilage and nuclei.
Form Liquid
For Use With (Application) Histology, Cytology
Label or Dye Dye
Melting Point ~0°C (32°F in water)
Molecular Formula Mixture
Packaging HDPE Bottle
pH Data not available.
Product Line Vion Histology
Purity 1%
Quantity 500 mL
Reagent Type Stain
Sample Type Human Cell Tissue
Solubility Complete in water.
Solution Type Aqueous
Storage Requirements Store in a cool, well-ventilated area away from incompatible substances.
Sub Cellular Localization Nuclei
Synonym Safranin O, Water Solution
Target Species Human
Target Specificity Cartilage
Technique Cell Staining
Product Type Safranin O
Assay 1.00%
Show More Show Less
ShelfLife 3 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.