Learn More
Volu Sol Safranin O Stain, 16 oz / 500 mL, Volu-Sol™
Supplier: Volu Sol VGC016
Safranin O, component of the Gram Stain Kit. General biological stain used in histology and cytology. Safranin colors cell nuclei red, making it a common counterstain in some staining protocols