Learn More
Electron Microscopy Sciences Safranin O, Certified, C.I. DcS-25 25 GR
SDP
Catalog No. 5028415
Encompass_Preferred
(Safranin T, Y or A; Basis Red 2; Cotton Red; Gossypimine)
C₂₀H₁₉N₄Cl F.W. 350.84
Solubility: Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% Xylene
Also known as Safranin T, Y or A, Basis Red 2, Cotton Red and Gossypimine.
Catalog No. 50-284-15 Supplier Electron Microscopy Sciences Supplier No. 20800
(Safranin T, Y or A; Basis Red 2; Cotton Red; Gossypimine)
C₂₀H₁₉N₄Cl F.W. 350.84
Solubility: Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% Xylene
Also known as Safranin T, Y or A, Basis Red 2, Cotton Red and Gossypimine.