Learn More
Safranin O, Certified, Sigma-Aldrich
Catalog No. 01259067 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
01-259-067 100 g
1 options
Catalog No. 01-259-067 Supplier MilliporeSigma Supelco Supplier No. S8884100G
Add to Cart
Add to Cart
Certified stain for chromosomes, Gram-negative bacteria, and plant cells.
Certified as a stain for chromosomes in the Flemming triple stain; to stain gram negative bacteria in the Gram stain procedure; Safranine-fast green FCF stain for plant cells.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Type Safranin O