Learn More
Sigma Aldrich Fine Chemicals Biosciences Safranin O for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0) | 477-73-6 | MFCD00011759 | 100G
Catalog No. 501806539
Encompass
Safranin O for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0) | Mol Wt: 350.84 | 477-73-6 | MFCD00011759 | 100G
Catalog No. 50-180-6539 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 84120100G
Safranin O for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0) | Mol Wt: 350.84 | 477-73-6 | MFCD00011759 | 100G