Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranin O, Sigma-Aldrich

Catalog No. 01258915 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-915 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-915 Supplier MilliporeSigma Supelco Supplier No. S2255100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cationic dye for histological staining and diagnostic assay manufacturing, with ≥85% dye content.

Safranin O has been used in the histological analysis of femurs. It has also been used to detect the presence of glycosaminoglycans in the inner body of meniscus structures.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥85 %
Quantity 100 g
Product Type Safranin O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.