Learn More
Electron Microscopy Sciences Safranin O Solution 0.1% in 0.1% Acetic Acid 500 ML
SDP
Catalog No. 5031979
Encompass_Preferred
Safranin O Solution in Acetic Acid is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.
Catalog No. 50-319-79 Supplier Electron Microscopy Sciences Supplier No. 2606905
Safranin O Solution in Acetic Acid is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.