Learn More
Abcam Safranin O, stain for Gram negative bacteria, 25G
Catalog No. 502704714
Encompass_Preferred
MW 350.84 g/mol. Biological stain used to stain Gram negative bacteria, chromosomes, mucin, cartilage and mast cells. Commonly used as a counter stain for gram-negative bacteria in the Gram stain procedure.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4714 Supplier Abcam Supplier No. AB14632925G
MW 350.84 g/mol. Biological stain used to stain Gram negative bacteria, chromosomes, mucin, cartilage and mast cells. Commonly used as a counter stain for gram-negative bacteria in the Gram stain procedure.
The product is subject to the following: Abcam Restricted Use Statement