Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranin T, Solstice

Catalog No. 60045874 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-874 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-874 Supplier Solstice Supplier No. 3103125G

missing translation for 'validMainframeMsg'

Redox indicator and for microscopy, S. No.: 967

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
PubChem CID 2723800
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 3924099
Quantity 25 g
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
Formula Weight 350.84g/mol
Chemical Name or Material Safranin T
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H318
EINECSNumber 207-518-8
RTECSNumber SG1623000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.