Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranine O, Ricca Chemical
SDP

Catalog No. RDCS005025F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS005025F 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS005025F Supplier Ricca Chemical Company Supplier No. RDCS005025F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
PubChem CID 2723800
IUPAC Name 2,7-diamino-1,8-dimethyl-5-phenyl-5λ5-phenazin-5-ylium chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Brown or Brown/Red to Green
Quantity 25 g
Grade Laboratory
Chemical Name or Material Safranine O
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.