Learn More
At Battery Company Inc SAFT LITHIUM BATTERY
SDP
Catalog No. NC0623364
Encompass_Preferred
SAFT LSH14 C 3.6V PRIMARY LITHIUM BATTERY The Saft LSH14 cylindrical primary lithium cells are based on Lithium-Thionyl chloride (Li-SOCl2) chemistry and feature high surface area spiral electrodes for maximum current capability. The LSH14 is designed for applications requiring continuous currents in the 0.1-1.8 A range, with superimposed pulses as high as 4 A. Construction features: Spiral electrodes Stainless steel container Hermetic and sturdy glass-to-metal sealing Built-in safety vents Individual cells with 5.5 A non-resettable fuse
Catalog No. NC0623364 Supplier At Battery Company Inc Supplier No. LSH14
SAFT LSH14 C 3.6V PRIMARY LITHIUM BATTERY The Saft LSH14 cylindrical primary lithium cells are based on Lithium-Thionyl chloride (Li-SOCl2) chemistry and feature high surface area spiral electrodes for maximum current capability. The LSH14 is designed for applications requiring continuous currents in the 0.1-1.8 A range, with superimposed pulses as high as 4 A. Construction features: Spiral electrodes Stainless steel container Hermetic and sturdy glass-to-metal sealing Built-in safety vents Individual cells with 5.5 A non-resettable fuse