Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SafTest™ Analyzer Liquid Sample Instrument Platform

Catalog No. MP07LI2020 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Application):
Glycerin, Malonaldehyde Testing
Peroxide, Alkenal Testing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
MP07LI2020 Peroxide, Alkenal Testing
MP07LI2000 Glycerin, Malonaldehyde Testing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07LI2020 Supplier MP Biomedicals™ Supplier No. 07LI2020
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

Timer, two block dri-bath, insert, vortex, test tube rack, pipette PDP, vacuum pump assembly, acrylic base, 100 to 1000 μL pipette PDP, 15 mm dri-block insert, 20 mm test tube rack, SafTest analyzer, tube rocker, O-ring for vacuum pump assembly

SafTest™ Analyzer Liquid Sample Instrument Platform is used in conjunction with alkenal, peroxide test kit.

  • Equipment platform for conducting tests
  • Contains all of instrumentation necessary to run solid samples
  • Test time: 30 minutes
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Dimensions: 30 x 27.2 x 12.5 cm
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Peroxide, Alkenal Testing
Quantity 1 Ea.
Storage Requirements Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.