Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SafTest™ Dispenser Set

Catalog No. MP7DSTG2000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Application):
Alkenal Testing
Free Fatty Acid Testing
Malonaldehyde Testing
Percent Fat Testing
Peroxide Testing
Total Glycerin Testing
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
MP7DSTG2000 Total Glycerin Testing
MP7DSAK2000 Alkenal Testing
MP7DSFA2000 Free Fatty Acid Testing
MP7DSPR2000 Peroxide Testing
MP7DSPF2000 Percent Fat Testing
MP7DSMA2000 Malonaldehyde Testing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP7DSTG2000 Supplier MP Biomedicals™ Supplier No. 07DSTG2000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack

Dispenser Set is an accessory for SafTest™ test kits.

  • Dispenser set for each chosen endpoint
  • Allows for accurate testing, every time
  • For use with SafTest™ kits
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Total Glycerin Testing
Storage Requirements Room temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.