Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SafTest™ Sample Consumables Platform

Catalog No. MP07LC2000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Sample Type:
Liquid Samples
Solid Samples
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
MP07LC2000 Liquid Samples
MP07SC2000 Solid Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07LC2000 Supplier MP Biomedicals™ Supplier No. 07LC2000
Add to Cart
Add to Cart

Includes

Printer paper, membranes, precision tissue wipers, solid glass beads, 15mL conical tubes, 10 to 100μL pipette tips, 200 to 1000μL pipette tips, 12mm culture tubes, 12mm culture tube caps, sharpie marke

SafTest™ Sample Consumables Platform contains all consumables necessary to run alkenal, peroxide tests.

  • Consumable platform for conducting alkenal, peroxide testing
  • Contains all of consumables necessary to run solid, liquid samples
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Peroxide, Alkenal Testing
Sample Type Liquid Samples
Quantity 1 Ea.
Storage Requirements Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.