Learn More
USA Scientific Inc Saf-T-Seal 2 mL Screw Cap Tube
Catalog No. 502744947
Encompass
Saf-T-Seal 2.0 mL Assembled Tethered-Screw Cap Tube, integrated o-ring, tethered white cap (not screwed on), graduated, self-standing base, clear, str, 100 assemblies/pack
Catalog No. 50-274-4947 Supplier USA Scientific Inc Supplier No. 14209410
Saf-T-Seal 2.0 mL Assembled Tethered-Screw Cap Tube, integrated o-ring, tethered white cap (not screwed on), graduated, self-standing base, clear, str, 100 assemblies/pack