Learn More
USA Scientific Inc Saf-T-Seal .5mL Screw Cap Tube
Catalog No. 502745235
Encompass
Saf-T-Seal 0.5mL Assembled Tethered-Screw Cap Tube, integrated o-ring, tethered white cap (not screwed on), self-standing base, clear, str, 100 assembles/pack.
Catalog No. 50-274-5235 Supplier USA Scientific Inc Supplier No. 14059410
Saf-T-Seal 0.5mL Assembled Tethered-Screw Cap Tube, integrated o-ring, tethered white cap (not screwed on), self-standing base, clear, str, 100 assembles/pack.