Learn More
MSC SAFTY SPLASH GOGGLE
Catalog No. 5000686436
Encompass
These safety goggles are designed for chemical splash protection and are suitable for various industrial applications. Featuring clear polycarbonate lenses with anti-fog and scratch-resistant coatings, they provide indirect venting and can fit comfortably over prescription eyewear.
  • Clear polycarbonate lenses with anti-fog and scratch-resistant coating
  • Indirect vent design for splash and dust protection
  • Universal size, designed to fit over most prescription eyewear
  • Adjustable elastic strap for a secure fit
  • Lenses absorb 99.9% of UV rays
  • Meets ANSI Z87.1+ standards
Catalog No. 50-006-86436 Supplier MSC Supplier No. 73409450
These safety goggles are designed for chemical splash protection and are suitable for various industrial applications. Featuring clear polycarbonate lenses with anti-fog and scratch-resistant coatings, they provide indirect venting and can fit comfortably over prescription eyewear.
  • Clear polycarbonate lenses with anti-fog and scratch-resistant coating
  • Indirect vent design for splash and dust protection
  • Universal size, designed to fit over most prescription eyewear
  • Adjustable elastic strap for a secure fit
  • Lenses absorb 99.9% of UV rays
  • Meets ANSI Z87.1+ standards