Learn More
AdipoGen SAG (1 mg)
SDP
Supplier:  AdipoGen AGCR13506M001
Encompass_Preferred
Chemical. CAS: 912545-86-9. Formula: C28H28ClN3OS. MW: 490.1. Cell permeable Sonic hedgehog (Shh) agonist. Cell permeable smoothened (Smo) agonist. Induces Sonic hedgehog (Shh) pathway activation and counteracts cyclopamine inhibition of Smo. Acts as an activator of Smo at low concentrations and as an inhibitor of Smo at very high concentrations. Potent activator of VEGF expression in vitro.
Catalog No. 50-205-3883