Learn More
Medchemexpress LLC HY-12848 1mg , SAG CAS:912545-86-9 Purity:>98%
SDP
Catalog No. 502259480
Encompass_Preferred
HY-12848 1mg SAG CAS: 912545-86-9 SAG is a potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM). SAG activates the Hedgehog signaling pathway and counteracts Cyclopamine (HY-17024) inhibition of Smo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9480 Supplier Medchemexpress LLC Supplier No. HY128481MG
HY-12848 1mg SAG CAS: 912545-86-9 SAG is a potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM). SAG activates the Hedgehog signaling pathway and counteracts Cyclopamine (HY-17024) inhibition of Smo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.