Learn More
AdipoGen SAG Analog (highly active) (1 mg)
SDP
Supplier:  AdipoGen AGCR13518M001
Encompass_Preferred
Chemical. CAS: 1401532-77-1. Formula: C28H27Cl2N3OS (cis-isomer). MW: 524.5. Cell permeable highly active Sonic hedgehog (Shh) agonist. Cell permeable smoothened (Smo) agonist.
Catalog No. 50-205-3900