Learn More
AdipoGen SAG Analog (LowTox) (1 mg)
SDP
Supplier:  AdipoGen AGCR13517M001
Encompass_Preferred
Chemical. CAS: 1401532-78-2. Formula: C28H27Cl2N3OS (trans-isomer). MW: 524.5. Cell permeable low toxicity Sonic hedgehog (Shh) agonist. Cell permeable smoothened (Smo) agonist. Potent activator of VEGF expression in vitro.
Catalog No. 50-205-3899