Learn More
TARGETMOL CHEMICALS INC SAG DIHYDROCHLORIDE 5MG
SDP
Catalog No. 502579986
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAG dihydrochloride is a potent Smoothened (Smo) receptor agonist ( EC 50 =3 nM; K d =59 nM). SAG dihydrochloride activates the Hedgehog signaling pathway and counteracts Cyclopamine inhibition of Smo. purity: 100%
Catalog No. 50-257-9986 Supplier TARGETMOL CHEMICALS INC Supplier No. T404655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAG dihydrochloride is a potent Smoothened (Smo) receptor agonist ( EC 50 =3 nM; K d =59 nM). SAG dihydrochloride activates the Hedgehog signaling pathway and counteracts Cyclopamine inhibition of Smo. purity: 100%