Learn More
Medchemexpress LLC HY-12848B 5mg Medchemexpress, SAG (hydrochloride) CAS:2095432-58-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12848B5MG
Encompass_Preferred
Medchemexpress, HY-12848B 5mg SAG (hydrochloride) CAS:2095432-58-7 SAG (hydrochloride) is a potent Smo receptor agonist, and activates the Hedgehog signaling pathway, with a K d of 59 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2622