Learn More
Medchemexpress LLC SAG Human P.pastor 50ug
SDP
Catalog No. 5000183416
Encompass_Preferred
Recombinant human SAG (Smoothened agonist) expressed in Pichia pastoris and purified with a histidine tag. Supplied as a purified protein for research use in hedgehog pathway activation and cell signaling assays; suitable for in vitro studies. Handle and store according to supplier recommendations to maintain activity and stability.
  • Recombinant human protein expressed in Pichia pastoris.
  • His affinity tag for facile purification.
  • Suitable for in vitro hedgehog pathway and signaling assays.
  • Supplied in microgram quantities for experimental use.
  • Store and handle according to supplier recommendations to preserve activity.
Catalog No. 50-001-83416 Supplier Medchemexpress LLC Supplier No. HYP7178150UG
Recombinant human SAG (Smoothened agonist) expressed in Pichia pastoris and purified with a histidine tag. Supplied as a purified protein for research use in hedgehog pathway activation and cell signaling assays; suitable for in vitro studies. Handle and store according to supplier recommendations to maintain activity and stability.
  • Recombinant human protein expressed in Pichia pastoris.
  • His affinity tag for facile purification.
  • Suitable for in vitro hedgehog pathway and signaling assays.
  • Supplied in microgram quantities for experimental use.
  • Store and handle according to supplier recommendations to preserve activity.