Learn More
Medchemexpress LLC SAG (Standard) | 912545-86-9 | 99.6% | 1 MG
SDP
Catalog No. 5000341849
Encompass_Preferred
This product serves as an analytical standard for a potent Smoothened (Smo) receptor agonist, which functions by activating the Hedgehog signaling pathway and counteracting the inhibitory effects of another compound on Smo.
  • Functions as an analytical standard.
  • Potent receptor agonist.
  • Activates a specific signaling pathway.
  • Counteracts inhibition of a receptor.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Commonly employed in HPLC, GC, and MS applications.
Catalog No. 50-003-41849 Supplier Medchemexpress LLC Supplier No. HY12848R1MG
This product serves as an analytical standard for a potent Smoothened (Smo) receptor agonist, which functions by activating the Hedgehog signaling pathway and counteracting the inhibitory effects of another compound on Smo.
  • Functions as an analytical standard.
  • Potent receptor agonist.
  • Activates a specific signaling pathway.
  • Counteracts inhibition of a receptor.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Commonly employed in HPLC, GC, and MS applications.