Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sage Science™ BluePippin™ 2% Agarose Gel Cassettes
SDP

Catalog No. 12100519
Encompass
Change view
Click to view available options
Quantity:
10 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-100-519 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-100-519 Supplier Sage Science™ Supplier No. BEF2010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For size selections between 100bp and 600bp. Sage Science™ BluePippin™ 2% Agarose Gel Cassettes are commonly used for Illumina paired-end library construction, Ion Torrent template prep, and ChIP applications.

  • Includes sufficient reagents to run all cassettes in pack
  • Dye free cassettes, with internal or external marker, runs four or five samples per cassette
  • Band capture cassettes contain dye and external marker, runs four samples per cassette

Specifications

Concentration 2%
Format Cassette
Separation Range 100 to 600 bp
Quantity 10 Pack
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.