Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sage Science™ BluePippin™ Dye Free 0.75% Agarose Gel Cassettes
SDP

Catalog No. 12100720
Encompass
Change view
Click to view available options
Quantity:
10 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-100-720 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-100-720 Supplier Sage Science™ Supplier No. BHZ7510
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sage Science™ BluePippin™ Dye Free 0.75% Agarose Gel Cassettes are disposable and pre-cast cassettes with agarose and running buffer.

  • Includes external markers
  • Uses marker Z1
  • Package includes sufficient marker and loading solution for 10 cassette
  • Sample capacity is 4 samples/cassette
  • Size range: 50kb target collection

Specifications

Concentration 0.75%
Format Cassette
Quantity 10 Pack
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.