Learn More
Lonza Walkersville SAGM Small Airway Epithelial Cell Growth Medium BulletKit
Catalog No. NC9501460
Encompass
SAGM BulletKit is small airway epithelial cell growth medium that is serum-free and has been optimized to grow human small airway epithelial cells (SAEC) from normal and diseased tissue.Content :1 x SABM Basal Medium (CC-3119), 500 mL1 x SAGM SingleQuots Supplement Pack (CC-4124) containing:1 x Orange Cap Vial with BPE, 2.0 mL1 x Lilac Cap Vial with Insulin, 0.5 mL1 x Natural Cap Vial with Hydrocortisone, 0.5 mL1 x Red Cap Vial with GA-1000, 0.5 mL1 x Amber Vial with Retinoic Acid, 0.5 mL1 x Bottle with Fatty Acid Free BSA, 5 mL1 x Natural Cap Vial with Transferrin, 0.5 mL1 x Amber Vial with Triiodothyronine, 0.5 mL1 x Amber Vial with Epinephrine, 0.5 mL1 x Green Cap Vial with hEGF, 0.5 mL
Catalog No. NC9501460 Supplier Lonza Walkersville Supplier No. CC3118
SAGM BulletKit is small airway epithelial cell growth medium that is serum-free and has been optimized to grow human small airway epithelial cells (SAEC) from normal and diseased tissue.Content :1 x SABM Basal Medium (CC-3119), 500 mL1 x SAGM SingleQuots Supplement Pack (CC-4124) containing:1 x Orange Cap Vial with BPE, 2.0 mL1 x Lilac Cap Vial with Insulin, 0.5 mL1 x Natural Cap Vial with Hydrocortisone, 0.5 mL1 x Red Cap Vial with GA-1000, 0.5 mL1 x Amber Vial with Retinoic Acid, 0.5 mL1 x Bottle with Fatty Acid Free BSA, 5 mL1 x Natural Cap Vial with Transferrin, 0.5 mL1 x Amber Vial with Triiodothyronine, 0.5 mL1 x Amber Vial with Epinephrine, 0.5 mL1 x Green Cap Vial with hEGF, 0.5 mL