Learn More
Lonza Walkersville SABM Small Airway Epithelial Cell Growth Basal Medium
Catalog No. NC9986192
Encompass
SABM is a basal medium sold separately. This is used as a component in the full SAGM Small Airway Epithelial Cell Growth Medium BulletKit (catalog number CC-3118) which consists of basal and supplements packaged together. SAGM BulletKit is serum-free and has been optimized to grow human small airway epithelial cells (SAEC).Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9986192 Supplier Lonza Walkersville Supplier No. CC3119
SABM is a basal medium sold separately. This is used as a component in the full SAGM Small Airway Epithelial Cell Growth Medium BulletKit (catalog number CC-3118) which consists of basal and supplements packaged together. SAGM BulletKit is serum-free and has been optimized to grow human small airway epithelial cells (SAEC).Content:1 x Bottle Basal Medium 500 mL