Learn More
Lonza Walkersville SAGM Small Airway Epithelial Cell Growth Medium SingleQuots Supplements and Growth Factors
Catalog No. NC0026372
Encompass
SAGM SingleQuot Kits are components sold separately and does not include the basal medium. These are part of the full SAGM Small Airway Epithelial Cell Growth Medium BulletKit (catalog number CC-3118) which consists of basal medium and supplements packaged together. SAGM BulletKit.SAGM BulletKit is serum-free and has been optimized to grow human small airway epithelial cells (SAEC).Content:1 x Orange Cap Vial with BPE 2.0 mL1 x Lilac Cap Vial with Insulin 0.5 mL1 x Natural Cap Vial with Hydrocortisone 0.5 mL1 x Red Cap Vial with GA-1000 0.5 mL1 x Amber Vial with Retinoic Acid 0.5 mL1 x Bottle with Fatty Acid Free BSA 5 mL1 x Natural Cap Vial with Transferrin 0.5 mL1 x Amber Vial with Triiodothyronine 0.5 mL1 x Amber Vial with Epinephrine 0.5 mL1 x Green Cap Vial with hEGF 0.5 mL
Catalog No. NC0026372 Supplier Lonza Walkersville Supplier No. CC4124
SAGM SingleQuot Kits are components sold separately and does not include the basal medium. These are part of the full SAGM Small Airway Epithelial Cell Growth Medium BulletKit (catalog number CC-3118) which consists of basal medium and supplements packaged together. SAGM BulletKit.SAGM BulletKit is serum-free and has been optimized to grow human small airway epithelial cells (SAEC).Content:1 x Orange Cap Vial with BPE 2.0 mL1 x Lilac Cap Vial with Insulin 0.5 mL1 x Natural Cap Vial with Hydrocortisone 0.5 mL1 x Red Cap Vial with GA-1000 0.5 mL1 x Amber Vial with Retinoic Acid 0.5 mL1 x Bottle with Fatty Acid Free BSA 5 mL1 x Natural Cap Vial with Transferrin 0.5 mL1 x Amber Vial with Triiodothyronine 0.5 mL1 x Amber Vial with Epinephrine 0.5 mL1 x Green Cap Vial with hEGF 0.5 mL