Learn More
Tocris™ SAH-SOS1A

Description
Sold under license from Dana-Farber Cancer Institute
Specifications
Specifications
| CAS | 1652561-87-9 |
| Content And Storage | Store at -20°C |
| Description | KRas/SOS1 interaction inhibitor |
| Formulation | C100H159N27O28 |
| Molecular Weight (g/mol) | 2187.53 |
| Quantity | 500 μg |
| Sequence | RRFFGIXLTNXLKTEEGN (Modifications: Arg-1 = N-terminal Ac, X = (S)-2-(4-pentenyl)alanine, X-7 and X-11 stapled together with a double bond) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.