Learn More
Medchemexpress LLC HY-111595 10mg Medchemexpress, SAHA chloroalkane T1 CAS:1613617-05-2 Purity:>98%
SDP
Catalog No. 501968923
Encompass_Preferred
Medchemexpress, HY-111595 10mg SAHA chloroalkane T1 CAS:1613617-05-2 SAHA chloroalkane T1 is a chloroalkane capture tag by tethering Vorinostat (SAHA) and a chloroalkane tag T1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8923 Supplier Medchemexpress LLC Supplier No. HY11159510MG
Medchemexpress, HY-111595 10mg SAHA chloroalkane T1 CAS:1613617-05-2 SAHA chloroalkane T1 is a chloroalkane capture tag by tethering Vorinostat (SAHA) and a chloroalkane tag T1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.