Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SAHARA S5P Heated PPO Bath

Catalog No. 13875171 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Water Bath
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
13-875-171 Water Bath
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-875-171 Supplier Thermo Scientific™ Supplier No. 1703050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

This is the bath only - not the full kit. The circulator is sold separately.

A polyphenylene oxide bath is an economical alternative to stainless steel

  • Bath volume: 3-5 L
  • Material: Polyphenylene oxide (PPO)
  • Thermally resistant up to 100°C
  • Temperature: Ambient plus 10°C to 100°C
TRUSTED_SUSTAINABILITY

Specifications

Includes Water Bath
Material Polyphenylene oxide (PPO)
Size Bath Volume: 3 to 5L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.