Learn More
Sarstedt Inc SAHARA-III MAXITHERM 115 V,
Supplier:  Sarstedt Inc 97.8710.802
Encompass_Preferred
for thawing and warming blood products in plastic bags before transfusion, max. capacity: 6 storage bags, suitable for plasma, whole blood, erythrocyte concentrates and cryopreserved preparations
Catalog No. NC2198291
Add to Cart
Request a Quote