Learn More
Medchemexpress LLC HY-126585 1mg , SAICAR CAS:3031-95-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1265851MG
Encompass_Preferred
HY-126585 1mg SAICAR CAS: 3031-95-6 SAICAR is an intermediate of de novo purine nucleotide biosynthesis, activates pyruvate kinase isoform M2 (PKM2) in an isozyme-selective manner, with an EC50 of 0.3 mM. SAICAR stimulates PKM2 and promotes cancer cell survival in glucose-limited conditions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9478