Learn More
TARGETMOL CHEMICALS INC Saikogenin A 1MG
SDP
Catalog No. 502877713
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Saikogenin A is a dipeptidyl peptidase-4 inhibitor. Purity 99.47%
Catalog No. 50-287-7713 Supplier TARGETMOL CHEMICALS INC Supplier No. T168381MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Saikogenin A is a dipeptidyl peptidase-4 inhibitor. Purity 99.47%