Learn More
Medchemexpress LLC HY-N6584 5mg Medchemexpress, Saikogenin A CAS:5092-09-1 Purity:>98%
SDP
Catalog No. 501933959
Encompass_Preferred
Medchemexpress, HY-N6584 5mg Saikogenin A CAS:5092-09-1 Saikogenin A, extracted from a Chinese herbal plant called Tsai-Fu, is a dipeptidyl peptidase-IV ( DPP-IV ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3959 Supplier Medchemexpress LLC Supplier No. HYN65845MG
Medchemexpress, HY-N6584 5mg Saikogenin A CAS:5092-09-1 Saikogenin A, extracted from a Chinese herbal plant called Tsai-Fu, is a dipeptidyl peptidase-IV ( DPP-IV ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.