Learn More
Medchemexpress LLC Saikogenin D | 5573-16-0 | 1 MG
SDP
Catalog No. 5000419353
Encompass_Preferred
Saikogenin D is isolated from Bupleurum chinense and possesses anti-inflammatory effects. It activates epoxygenases, converting arachidonic acid to epoxyeicosanoids and dihydroxyeicosatrienoic acids, which then secondarily inhibit prostaglandin E2 (PGE2) production. It also leads to an elevation of intracellular calcium ([Ca2+]i) due to Ca2+ release from intracellular stores.
  • Inhibits PGE2 production in C6 rat glioma cells
  • Elevates intracellular calcium in C6 rat glioma cells
Catalog No. 50-004-19353 Supplier Medchemexpress LLC Supplier No. HYN42371MG
Saikogenin D is isolated from Bupleurum chinense and possesses anti-inflammatory effects. It activates epoxygenases, converting arachidonic acid to epoxyeicosanoids and dihydroxyeicosatrienoic acids, which then secondarily inhibit prostaglandin E2 (PGE2) production. It also leads to an elevation of intracellular calcium ([Ca2+]i) due to Ca2+ release from intracellular stores.
  • Inhibits PGE2 production in C6 rat glioma cells
  • Elevates intracellular calcium in C6 rat glioma cells