Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikogenin D | 5573-16-0 | 5 MG
SDP

Catalog No. 5000420299
Encompass_Preferred

Saikogenin D is a compound isolated from Bupleurum chinense with anti-inflammatory effects. It activates epoxygenases, which convert arachidonic acid into epoxyeicosanoids and dihydroxyeicosatrienoic acids. These metabolites, in turn, inhibit prostaglandin E2 (PGE2) production. Additionally, Saikogenin D leads to an elevation of intracellular Ca2+ ([Ca2+]i) due to Ca2+ release from intracellular stores.

  • Purity: 99.99%
  • Molecular weight: 472.70
  • Formula: C30H48O4
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants (Compositae Carphephorus corymbosus (Nutt.) Torr. & A.Gray, Umbelliferae)

Catalog No. 50-004-20299 Supplier Medchemexpress LLC Supplier No. HYN42375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikogenin D is a compound isolated from Bupleurum chinense with anti-inflammatory effects. It activates epoxygenases, which convert arachidonic acid into epoxyeicosanoids and dihydroxyeicosatrienoic acids. These metabolites, in turn, inhibit prostaglandin E2 (PGE2) production. Additionally, Saikogenin D leads to an elevation of intracellular Ca2+ ([Ca2+]i) due to Ca2+ release from intracellular stores.

  • Purity: 99.99%
  • Molecular weight: 472.70
  • Formula: C30H48O4
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants (Compositae Carphephorus corymbosus (Nutt.) Torr. & A.Gray, Umbelliferae)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.