Learn More
Medchemexpress LLC Saikogenin F | 14356-59-3 | 1 MG
SDP
Catalog No. 5000417516
Encompass_Preferred
Saikogenin F is a chemical compound known for its anti-cancer activity. It has been observed to inhibit the proliferation of various cancer cell lines, demonstrating its potential in research applications.
  • Inhibits the proliferation of A549 cells with increasing rates at concentrations of 50, 70, and 90 μg/mL.
  • Shows cytotoxicity against human A549, Bcap37, Hep 3B2, HepG2, and MCF7 cells in vitro.
Catalog No. 50-004-17516 Supplier Medchemexpress LLC Supplier No. HYN76701MG
Saikogenin F is a chemical compound known for its anti-cancer activity. It has been observed to inhibit the proliferation of various cancer cell lines, demonstrating its potential in research applications.
  • Inhibits the proliferation of A549 cells with increasing rates at concentrations of 50, 70, and 90 μg/mL.
  • Shows cytotoxicity against human A549, Bcap37, Hep 3B2, HepG2, and MCF7 cells in vitro.