Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin A | 20736-09-8 | 99.3% | 780.98 g/mol | C42H68O13 | 10 MG
SDP

Catalog No. 5000296619
Encompass_Preferred

Saikosaponin A is a triterpenoid saponin research standard used in biological and pharmacological studies for its anti-inflammatory, neuroprotective, and lipid-metabolism effects. It is supplied as a solid suitable for in vitro and in vivo formulation.

  • Chemical formula: C42H68O13.
  • Molecular weight: 780.98 g/mol.
  • Cas number: 20736-09-8.
  • Purity: 99.3% (LCMS).
  • Solubility: DMSO 100 mg/mL; additional in vivo formulations described in supplier data.
  • Storage: store at -20°C and protect from light; in solvent, -80°C up to 6 months.
  • Pack size: 10 mg, supplied as a solid.

Catalog No. 50-002-96619 Supplier Medchemexpress LLC Supplier No. HYN024610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Saikosaponin A is a triterpenoid saponin research standard used in biological and pharmacological studies for its anti-inflammatory, neuroprotective, and lipid-metabolism effects. It is supplied as a solid suitable for in vitro and in vivo formulation.

  • Chemical formula: C42H68O13.
  • Molecular weight: 780.98 g/mol.
  • Cas number: 20736-09-8.
  • Purity: 99.3% (LCMS).
  • Solubility: DMSO 100 mg/mL; additional in vivo formulations described in supplier data.
  • Storage: store at -20°C and protect from light; in solvent, -80°C up to 6 months.
  • Pack size: 10 mg, supplied as a solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.