Learn More
Cayman Chemical Saikosaponin B1, Purity: ≥98%, CAS: 58558-08-0, 25 mg
SDP
Catalog No. 502948416
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; an agonist of the 5-HT2C receptor (EC50 = 147.41 µM in HEK293 cells expressing the human receptor in a calcium mobilization assay); stimulates PGE2 production in isolated rat peritoneal macrophages at 100 µg/ml; protects SH-SY5Y cells from hydrogen peroxide-induced cell death at 50 µg/ml
Catalog No. 50-294-8416 Supplier Cayman Chemical Supplier No. 3460925 MG
A triterpenoid saponin with diverse biological activities; an agonist of the 5-HT2C receptor (EC50 = 147.41 µM in HEK293 cells expressing the human receptor in a calcium mobilization assay); stimulates PGE2 production in isolated rat peritoneal macrophages at 100 µg/ml; protects SH-SY5Y cells from hydrogen peroxide-induced cell death at 50 µg/ml