Learn More
TARGETMOL CHEMICALS INC Saikosaponin B1 5MG
SDP
Catalog No. 502876112
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Saponin activator. Activates release of Prostaglandin E2 in vitro. Anti-inflammatory agent. Orally active. Active in vivo and in vitro. Purity 99.7%
Catalog No. 50-287-6112 Supplier TARGETMOL CHEMICALS INC Supplier No. T39125MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Saponin activator. Activates release of Prostaglandin E2 in vitro. Anti-inflammatory agent. Orally active. Active in vivo and in vitro. Purity 99.7%