Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin B2 | 58316-41-9 | MFCD00210366 | 99.5% | 780.98 | C42H68O13 | 10 MG
SDP

Catalog No. 5000296307
Encompass_Preferred

Saikosaponin B2 is a natural triterpenoid saponin isolated from Bupleurum chinense root. It is reported to act as an entry inhibitor against hepatitis C virus (HCV) and has documented anti-cancer activity. Supplied for research use, the compound is accompanied by analytical documentation and solubility guidance.

  • High purity - 99.5%.
  • Molecular formula C42H68O13.
  • Molecular weight 780.98.
  • Soluble in DMSO; in vitro solubility reported at 100 mg/mL (ultrasonic recommended).
  • Includes SDS, COA, HNMR, and LCMS documentation.
  • Available in small quantities suitable for analytical or biological research (10 MG).

Catalog No. 50-002-96307 Supplier Medchemexpress LLC Supplier No. HYN024810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin B2 is a natural triterpenoid saponin isolated from Bupleurum chinense root. It is reported to act as an entry inhibitor against hepatitis C virus (HCV) and has documented anti-cancer activity. Supplied for research use, the compound is accompanied by analytical documentation and solubility guidance.

  • High purity - 99.5%.
  • Molecular formula C42H68O13.
  • Molecular weight 780.98.
  • Soluble in DMSO; in vitro solubility reported at 100 mg/mL (ultrasonic recommended).
  • Includes SDS, COA, HNMR, and LCMS documentation.
  • Available in small quantities suitable for analytical or biological research (10 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.