Learn More
Medchemexpress LLC Saikosaponin B3 | 58316-42-0 | 5 MG
SDP
Catalog No. 5000420440
Encompass_Preferred
Saikosaponin B3 is a triterpene saikosaponin isolated from the roots of *Bupleurum falcatum* L. This off-white to light yellow solid has a molecular weight of 813.02 and a chemical formula of C43H72O14. It is intended for research use only.
  • Isolated from *Bupleurum falcatum* L.
  • Exhibits an analgesic effect
  • Inhibits ACTH-induced lipolysis in fat cells
  • Solid appearance, off-white to light yellow
Catalog No. 50-004-20440 Supplier Medchemexpress LLC Supplier No. HYN42195MG
Saikosaponin B3 is a triterpene saikosaponin isolated from the roots of *Bupleurum falcatum* L. This off-white to light yellow solid has a molecular weight of 813.02 and a chemical formula of C43H72O14. It is intended for research use only.
  • Isolated from *Bupleurum falcatum* L.
  • Exhibits an analgesic effect
  • Inhibits ACTH-induced lipolysis in fat cells
  • Solid appearance, off-white to light yellow