Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin B4 | 58558-09-1 | 99.0% | 813.02 | C43H72O14 | 10 MG
SDP

Catalog No. 5000289692
Encompass_Preferred

Saikosaponin B4 is a triterpenoid saponin isolated from the roots of Bupleurum chinensis. It is used in biochemical and pharmacological research and has been reported to selectively inhibit ACTH-induced lipolysis.

  • Triterpenoid saponin isolated from Bupleurum chinensis.
  • Selectively inhibits ACTH-induced lipolysis.
  • High purity (99.04%) suitable for research applications.
  • Molecular formula C43H72O14; molecular weight 813.02.
  • CAS number 58558-09-1 for clear identification.
  • Store protected from light at 4°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-89692 Supplier Medchemexpress LLC Supplier No. HYN421810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin B4 is a triterpenoid saponin isolated from the roots of Bupleurum chinensis. It is used in biochemical and pharmacological research and has been reported to selectively inhibit ACTH-induced lipolysis.

  • Triterpenoid saponin isolated from Bupleurum chinensis.
  • Selectively inhibits ACTH-induced lipolysis.
  • High purity (99.04%) suitable for research applications.
  • Molecular formula C43H72O14; molecular weight 813.02.
  • CAS number 58558-09-1 for clear identification.
  • Store protected from light at 4°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.