Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin B4 | 58558-09-1 | MFCD30725477 | 5 MG
SDP

Catalog No. 5000419357
Encompass_Preferred

Saikosaponin B4 is a chemical compound with the molecular formula C43H72O14 and a molecular weight of 813.02. It is typically a white to yellow solid, with a high purity of 99.72% as determined by HPLC.

  • White to yellow solid appearance
  • Purity (HPLC) of 99.72%
  • Consistent 1H NMR spectrum
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-19357 Supplier Medchemexpress LLC Supplier No. HYN42185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin B4 is a chemical compound with the molecular formula C43H72O14 and a molecular weight of 813.02. It is typically a white to yellow solid, with a high purity of 99.72% as determined by HPLC.

  • White to yellow solid appearance
  • Purity (HPLC) of 99.72%
  • Consistent 1H NMR spectrum
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.