Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin C | 20736-08-7 | 98.1% | 927.12 g/mol | C48H78O17 | 10 MG
SDP

Catalog No. 5000296394
Encompass_Preferred

Saikosaponin C is a natural triterpene saponin used as a research reagent for studies of antiviral, anti-inflammatory, and neuronal biology. It is supplied as a high-purity analytical standard for in vitro biochemical and cellular assays.

  • natural triterpene saponin isolated from Radix Bupleuri.
  • used for in vitro research into antiviral and anti-inflammatory pathways.
  • purity 98.09% as provided by supplier.
  • molecular weight 927.12 g/mol and CAS RN 20736-08-7.
  • commonly available in small research sizes such as 5 mg, 10 mg, and 20 mg.

Catalog No. 50-002-96394 Supplier Medchemexpress LLC Supplier No. HYN024910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin C is a natural triterpene saponin used as a research reagent for studies of antiviral, anti-inflammatory, and neuronal biology. It is supplied as a high-purity analytical standard for in vitro biochemical and cellular assays.

  • natural triterpene saponin isolated from Radix Bupleuri.
  • used for in vitro research into antiviral and anti-inflammatory pathways.
  • purity 98.09% as provided by supplier.
  • molecular weight 927.12 g/mol and CAS RN 20736-08-7.
  • commonly available in small research sizes such as 5 mg, 10 mg, and 20 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.