Learn More
TARGETMOL CHEMICALS INC SAIKOSAPONIN D 25MG
SDP
Catalog No. 502580066
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Saikosaponin D is a novel SERCA inhibitor by inhibiting NF-(kappa)B and STAT3 signaling to protect against acetaminophen-induced hepatotoxicity. purity: 99%
Catalog No. 50-258-0066 Supplier TARGETMOL CHEMICALS INC Supplier No. T391325MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Saikosaponin D is a novel SERCA inhibitor by inhibiting NF-(kappa)B and STAT3 signaling to protect against acetaminophen-induced hepatotoxicity. purity: 99%