Learn More
Selleck Chemical LLC Saikosaponin D S5454-5mg
SDP
Catalog No. 503108595
Encompass_Preferred
Saikosaponin D a calcium mobilizing agent (SERCA inhibitor) is a type of Saponin derivative which is a component extracted from Bupleurum falactum Saikosaponin D has anti-cancer activities
Catalog No. 50-310-8595 Supplier Selleck Chemical LLC Supplier No. S54545MG
Saikosaponin D a calcium mobilizing agent (SERCA inhibitor) is a type of Saponin derivative which is a component extracted from Bupleurum falactum Saikosaponin D has anti-cancer activities